Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Adtl-SA1215 | 782387-91-1 | 99.0% | 657.32 | C26H29I2NO3 | 5 MG
SDP

Catalog No. 5000221752
Encompass_Preferred

ADTL-SA1215 is a small-molecule activator of SIRT3 used in preclinical research to modulate autophagy in triple-negative breast cancer models. It is provided as a purified solid and as a DMSO solution, and includes reported solubility, storage, and in vivo formulation guidance for laboratory use.

  • Activates SIRT3 and modulates autophagy in cancer models.
  • High reported purity (≈98.96%).
  • Available as solid and as a 10 mM solution in DMSO.
  • Soluble in DMSO up to 100 mg/mL with ultrasonic warming.
  • In vivo formulation examples provided for multiple vehicles.
  • Recommended storage temperatures for powder and solutions are listed.
  • Molecular formula C26H29I2NO3; molecular weight 657.32.

Catalog No. 50-002-21752 Supplier Medchemexpress LLC Supplier No. HY1397425MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ADTL-SA1215 is a small-molecule activator of SIRT3 used in preclinical research to modulate autophagy in triple-negative breast cancer models. It is provided as a purified solid and as a DMSO solution, and includes reported solubility, storage, and in vivo formulation guidance for laboratory use.

  • Activates SIRT3 and modulates autophagy in cancer models.
  • High reported purity (≈98.96%).
  • Available as solid and as a 10 mM solution in DMSO.
  • Soluble in DMSO up to 100 mg/mL with ultrasonic warming.
  • In vivo formulation examples provided for multiple vehicles.
  • Recommended storage temperatures for powder and solutions are listed.
  • Molecular formula C26H29I2NO3; molecular weight 657.32.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.