Learn More
Medchemexpress LLC Adtn hydrobromide | 13575-86-5 | 98.0% | 10 MG
SDP
Catalog No. 5000232488
Encompass_Preferred
ADTN hydrobromide is the hydrobromide salt of ADTN, a long-acting dopamine receptor agonist used in research to probe dopaminergic signaling. It is supplied as a small, high-purity reagent for laboratory studies and has been reported to reduce behavioral visual threshold in DA-IPC-depleted zebrafish.
  • Long-acting dopamine receptor agonist.
  • Used to study dopaminergic signaling and zebrafish visual behavior.
  • High purity: 98.0%.
  • Supplied in a small research pack suitable for small-scale assays.
  • Molecular formula C10H14BrNO2, molecular weight 260.13 g/mol.
Catalog No. 50-002-32488 Supplier Medchemexpress LLC Supplier No. HY121650A10MG
ADTN hydrobromide is the hydrobromide salt of ADTN, a long-acting dopamine receptor agonist used in research to probe dopaminergic signaling. It is supplied as a small, high-purity reagent for laboratory studies and has been reported to reduce behavioral visual threshold in DA-IPC-depleted zebrafish.
  • Long-acting dopamine receptor agonist.
  • Used to study dopaminergic signaling and zebrafish visual behavior.
  • High purity: 98.0%.
  • Supplied in a small research pack suitable for small-scale assays.
  • Molecular formula C10H14BrNO2, molecular weight 260.13 g/mol.