Learn More
Medchemexpress LLC HY-12885B 1mg Medchemexpress, ADU-S100 (ammonium salt) CAS:1638750-96-5 Purity:>98%
SDP
Catalog No. 502028930
Encompass_Preferred
Medchemexpress, HY-12885B 1mg ADU-S100 (ammonium salt) CAS:1638750-96-5 ADU-S100 ammonium salt (MIW815 ammonium salt), an activator of stimulator of interferon genes (STING), leads to potent and systemic tumor regression and immunity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8930 Supplier Medchemexpress LLC Supplier No. HY12885B1MG
Add to Cart
Add to Cart
Medchemexpress, HY-12885B 1mg ADU-S100 (ammonium salt) CAS:1638750-96-5 ADU-S100 ammonium salt (MIW815 ammonium salt), an activator of stimulator of interferon genes (STING), leads to potent and systemic tumor regression and immunity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.