Learn More
Medchemexpress LLC ADU-S100 AMMONIUM SALT (5MG)
SDP
Catalog No. NC1622478
Encompass_Preferred
CAS 1638750-96-5 purity 99.23%,ADU-S100 ammonium salt is an activator of stimulator of interferon genes (STING).
Catalog No. NC1622478 Supplier Medchemexpress LLC Supplier No. HY12885B5MG
CAS 1638750-96-5 purity 99.23%,ADU-S100 ammonium salt is an activator of stimulator of interferon genes (STING).