Learn More
Medchemexpress LLC HY-12885C 5mg Medchemexpress, ADU-S100 enantiomer (Ammonium salt) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12885C5MG
Encompass_Preferred
Medchemexpress, HY-12885C 5mg ADU-S100 enantiomer (Ammonium salt) CAS: ADU-S100 enantiomer (Ammonium salt) is the less active enantiomer of ADU-S100. ADU-S100 is an activator of stimulator of interferon genes (STING) [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2641