Learn More
Cf Medical Inc Adult Smar pads Cartridge
SDP
Catalog No. NC1631221
Encompass_Preferred
Designed to be used in the HeartStart OnSite (HS1) AED only. The pads are “SMART” because they work with the device to sense whether they have been removed from the cartridge and placed on the patient’s skin. Pictures on the pads illustrate correct placement.
This item is not returnable. View return policy
Catalog No. NC1631221 Supplier Cf Medical Inc Supplier No. M5071A
Designed to be used in the HeartStart OnSite (HS1) AED only. The pads are “SMART” because they work with the device to sense whether they have been removed from the cartridge and placed on the patient’s skin. Pictures on the pads illustrate correct placement.