Learn More
ATCC Adult Melanocyte Growth Kit
Catalog No. 502382275
Encompass
Adult Melanocyte Growth Kit contains components that when added to Dermal Cell Basal Medium (ATCC PCS-200-030) create a complete ATCC Primary Cell Solutions culture environment for melanocytes derived from a normal adult human epidermis (eg, Primary Epidermal Melanocytes; Normal, Human, Adult, ATCC PCS-200-013).
Catalog No. 50-238-2275 Supplier ATCC Supplier No. PCS200042
Adult Melanocyte Growth Kit contains components that when added to Dermal Cell Basal Medium (ATCC PCS-200-030) create a complete ATCC Primary Cell Solutions culture environment for melanocytes derived from a normal adult human epidermis (eg, Primary Epidermal Melanocytes; Normal, Human, Adult, ATCC PCS-200-013).