Learn More
Med Vet International Adult Universal "F" Anesthesia Circuit, 2 Liter Bag, Each
Catalog No. 5000582547
Encompass_Preferred
This is the popular uni-limb circuit that is a tube within a tube circuit. Includes one 40" hose for lighter weight and maneuverability around patients head. Exhaled gas warms up incoming fresh gas. Attaches to any standard anesthesia machine.
Catalog No. 50-005-82547 Supplier Med Vet International Supplier No. JOR248F
This is the popular uni-limb circuit that is a tube within a tube circuit. Includes one 40" hose for lighter weight and maneuverability around patients head. Exhaled gas warms up incoming fresh gas. Attaches to any standard anesthesia machine.