Learn More
Cliawaived Inc UrineCHECK 7 Adulterant Tests
SDP
Catalog No. NC1859622
Encompass_Preferred
(25 Strips Per Bottle)The UrineCHECK 7 includes assays for Creatinine, pH, Specific Gravity, Nitrite, Bleach, Pyridinium Chlorochromate and Glutaraldehyde. The test strip is used to determine specimen (urine) integrity and is for forensic and use only. The UrineCheck 7 adulterant strips are not for use in diagnosing disease or illness to effect treatment, and/or cure. For in vitro use only.NOTE: This product is not CLIA Waived. This test is for FORENSIC USE ONLY and not for medical office use.
This item is not returnable. View return policy
Catalog No. NC1859622 Supplier Cliawaived Inc Supplier No. TCURINECHECK7
(25 Strips Per Bottle)The UrineCHECK 7 includes assays for Creatinine, pH, Specific Gravity, Nitrite, Bleach, Pyridinium Chlorochromate and Glutaraldehyde. The test strip is used to determine specimen (urine) integrity and is for forensic and use only. The UrineCheck 7 adulterant strips are not for use in diagnosing disease or illness to effect treatment, and/or cure. For in vitro use only.NOTE: This product is not CLIA Waived. This test is for FORENSIC USE ONLY and not for medical office use.