Learn More
MSC Lysol All-Purpose Cleaner, Lemon Breeze Scent, Spray Bottle, 32 oz
Catalog No. 502867114
Encompass
This advanced formulation cleaner delivers fresh, deep-cleaned, and disinfected surfaces by powering through tough cleaning jobs, cutting through grease and soap scum.
  • Eliminates 99.9% of viruses and bacteria when used as directed
  • Effective against SARS-CoV-2 on hard, non-porous surfaces in 2 minutes
  • Cuts through tough grease and soap scum
Catalog No. 50-286-7114 Supplier MSC Supplier No. 28652311
This advanced formulation cleaner delivers fresh, deep-cleaned, and disinfected surfaces by powering through tough cleaning jobs, cutting through grease and soap scum.
  • Eliminates 99.9% of viruses and bacteria when used as directed
  • Effective against SARS-CoV-2 on hard, non-porous surfaces in 2 minutes
  • Cuts through tough grease and soap scum