Learn More
Med Vet International Advantage Multi For Cats Purple, 9.1-18Lbs, 6 Pack
Catalog No. 502828768
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Advantage Multi for Cats (imidacloprid + moxidectin)Topical Solution combines 100 percent prevention of heartworm disease, as shown in clinical trials, with the flea control of Advantage, and the treatment and control of intestinal worms (hookworm and roundworm) - all in one, convenient, monthly topical solution. Advantage Multi for Cats also treats and controls ear mite infestations.
Catalog No. 50-282-8768 Supplier Med Vet International Supplier No. RXADVMULTP
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Advantage Multi for Cats (imidacloprid + moxidectin)Topical Solution combines 100 percent prevention of heartworm disease, as shown in clinical trials, with the flea control of Advantage, and the treatment and control of intestinal worms (hookworm and roundworm) - all in one, convenient, monthly topical solution. Advantage Multi for Cats also treats and controls ear mite infestations.