Learn More
Electron Microscopy Sciences Advanced Protocol Incubator Model 51028068, 120 V, 178 L chamber
SDP
Catalog No. 501902390
Encompass_Preferred
Dual convection for application versatility-fan speed adjustable from 0 to 100%.
Internal socket allows the use of equipment, such as shakers or stirrers, inside the unit.
Easy to clean, corrosion-resistant stainless-steel interior (AISI 304).
Broad temperature range from 5 degree C above ambient to 105 degree C-suitable for drying applications.
Temperature uniformity of +/- 0.6 (Fan off) and between +/-0.2 degree C and +/- 0.4 degree C (Fan full speed) (measured at 37 degree C).
Temperature stability of +/- 0.1 degree C (at 37 degree C).
Intuitive user interface for setting temperature.
Large, easy to read vacuum fluorescent display.
Internal glass door allows sample viewing without impacting temperature. Exceptionally small footprint. Convection Technology: Dual Convection.
Chamber: 178 L
Catalog No. 50-190-2390 Supplier Electron Microscopy Sciences Supplier No. 6315430
Add to Cart
Add to Cart
Request a Quote
Dual convection for application versatility-fan speed adjustable from 0 to 100%.
Internal socket allows the use of equipment, such as shakers or stirrers, inside the unit.
Easy to clean, corrosion-resistant stainless-steel interior (AISI 304).
Broad temperature range from 5 degree C above ambient to 105 degree C-suitable for drying applications.
Temperature uniformity of +/- 0.6 (Fan off) and between +/-0.2 degree C and +/- 0.4 degree C (Fan full speed) (measured at 37 degree C).
Temperature stability of +/- 0.1 degree C (at 37 degree C).
Intuitive user interface for setting temperature.
Large, easy to read vacuum fluorescent display.
Internal glass door allows sample viewing without impacting temperature. Exceptionally small footprint. Convection Technology: Dual Convection.
Chamber: 178 L