Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Advance Tabco™ Premium Series Stainless-Steel Work Tables

Catalog No. 19033551A Shop All Advance Tabco Products
Change view
Click to view available options
Length (Metric):
9.1 mm
76.2 cm
91.44 cm
121.92 cm
152.4 cm
182.88 cm
243 cm
Width (Metric):
60.96 cm
76.2 cm
Includes:
Three hat channels
Two hat channels
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Width (Metric) Includes
19-033-551A 76.2 cm 60.96 cm Two hat channels
19-033-551C 91.44 cm 60.96 cm Two hat channels
19-033-551D 121.92 cm 60.96 cm Two hat channels
19-03-3-551E 152.4 cm 60.96 cm Two hat channels
19-033-551F 182.88 cm 60.96 cm Two hat channels
19-033-552E 152.4 cm 76.2 cm Three hat channels
19-033-552D 121.92 cm 76.2 cm Three hat channels
19-033-552F 182.88 cm 76.2 cm Three hat channels
19-999-597 - - -
19-999-596 76.2 cm 60.96 cm Two hat channels
19-999-601 182.88 cm 60.96 cm Two hat channels
19-033-554D 121.92 cm 76.2 cm Three hat channels
19-033-554E 152.4 cm 76.2 cm Three hat channels
19-033-554G - - -
19-033-554F 182.88 cm 76.2 cm Three hat channels
19-999-595 243 cm 60.96 cm Two hat channels
19-999-598 9.1 mm 76.2 cm Two hat channels
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
Catalog No. 19-033-551A Supplier Advance Tabco™ Supplier No. SS240
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Wide, adjustable, and with a turned up edge for wall protection, the Advance Tabco Stainless-Steel Work Table is made for, and can take, heavy use in nearly all environments.

  • Top is furnished with 4.12cm (1.625 in.) sanitary rolled rim edges on front and back and square side edges
  • To reinforce and maintain level working surface, tables are supplied with hat channels
  • Pre-engineered welded angle adapters insure ease of future drawer installation

Model Specific:

SS - Series

  • Aluminum die cast leg-to-shelf clamp secures shelf to leg eliminating unsightly nuts and bolts. Undershelf is adjustable
  • Top: 14 gauge stainless steel type 304 series
  • Shelf: 18 gauge stainless steel
  • Legs: 4.12cm (1.625 in.) diameter tubular stainless steel. 2.54cm (1 in.) adjustable stainless steel bullet feet. Stainless steel gussets

TSS - Series

  • Front to back Stretchers are welded to legs. Left to right
  • Top: 14 gauge stainless steel type 304 series
  • Stretchers: 4.12cm (1.625 in.) dia. tubular stainless steel. LEGS: 4.12cm (1.625 in.) dia. tubular stainless steel type 304 series
  • 2.54cm (1 in.) adjustable stainless steel bullet feet. Stainless steel gussets. Stretcher bolted to legs and requires assembly

Specifications

Height (English) 35.5 in.
Length (English) 30 in.
Weight (Metric) 22.2 kg
Width (English) 24 in.
Certifications/Compliance NSF
Includes Two hat channels
Height (Metric) 90.17 cm
Length (Metric) 76.2 cm
Weight (English) 49 lb.
Width (Metric) 60.96 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.