Learn More
Agilent Technologies ADVANCEBIO GLY-X DEGLYCOSYLATI
Catalog No. NC2179415
Encompass
AdvanceBio Gly-X deglycosylation module, 96-ct (formerly ProZyme). Utilizes a novel 5- minute in-solution enzymatic protein deglycosylation (AdvanceBio N-glycanase, PNGaseF).
This item is not returnable. View return policy
Catalog No. NC2179415 Supplier Agilent Technologies Supplier No. GX96100
AdvanceBio Gly-X deglycosylation module, 96-ct (formerly ProZyme). Utilizes a novel 5- minute in-solution enzymatic protein deglycosylation (AdvanceBio N-glycanase, PNGaseF).