Learn More
Agilent Technologies ADVANCEBIO SEC 200A 1.9UM 2.1X
Catalog No. NC1927948
Encompass
AdvanceBio SEC 200A 1.9um 2.1x150mm PEEK
Catalog No. NC1927948 Supplier Agilent Technologies Supplier No. PL19803201PK
AdvanceBio SEC 200A 1.9um 2.1x150mm PEEK