Learn More
Agilent Technologies AdvanceBio Spin Columns, Analytical-Scale, Cross-Linked Dextran, 25/Pk.
Catalog No. NC3782400
Encompass
These spin columns are designed for efficient sample preparation of proteins and oligonucleotides under aqueous conditions, effectively removing excess salt and small molecule matrix components that can interfere with analysis. Using a centrifuge-based approach, these columns provide high-throughput desalting and buffer exchange while preserving native biomolecular structure.
  • Effective salt removal of 99.6%
  • Suitable for proteins >5 kDa and oligonucleotides >10 bp/nt
  • Sample preparation completed in 10 to 15 minutes
  • Compatible with a wide range of common buffers and denaturing agents
  • Stable across a pH range of 2.0 to 13.0
  • Includes 1.5 mL microcentrifuge collection tubes
  • Scalable for various sample numbers and volumes
Catalog No. NC3782400 Supplier Agilent Technologies Supplier No. 19801103
These spin columns are designed for efficient sample preparation of proteins and oligonucleotides under aqueous conditions, effectively removing excess salt and small molecule matrix components that can interfere with analysis. Using a centrifuge-based approach, these columns provide high-throughput desalting and buffer exchange while preserving native biomolecular structure.
  • Effective salt removal of 99.6%
  • Suitable for proteins >5 kDa and oligonucleotides >10 bp/nt
  • Sample preparation completed in 10 to 15 minutes
  • Compatible with a wide range of common buffers and denaturing agents
  • Stable across a pH range of 2.0 to 13.0
  • Includes 1.5 mL microcentrifuge collection tubes
  • Scalable for various sample numbers and volumes