Learn More
StatLab Advanced Carbol Fuchsin
SDP
Catalog No. 22021392
Change view
Click to view available options
Volume (Metric):
500 mL
Catalog No. Volume (Metric)
22-021-392 500 mL
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-021-392 Supplier StatLab Supplier No. STACFPT
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
StatLab Advanced Carbol Fuchsin - a 10-minute acid fast bacteria stain
  • It is used in the Ziehl-Neelsen stain, a differential stain, to detect acid-fast bacteria like Mycobacterium tuberculosis
  • Carbol Fuchsin is more soluble in the cells' wall lipids than in the acid alcohol, allowing the bacteria to retain the red color even after decolorization with acid alcohol
  • This staining method is crucial for differentiating acid-fast bacteria from non-acid-fast bacteria, which are easily decolorized
  • A MasterTech Special Stain Kit Component
Specifications
Volume (Metric) 500 mL
Sterility Non-sterile