Learn More
MURATA VIOS INC Advanced Cell Strainer CELLNETTA 5µm Case of 1
Catalog No. NC3918263
Encompass
New approach to cell separation by size. Ideal for a wide range of cell processing applications, including fractionation, concentration, and filtration.
Highlights:
・Fast and simple processing
・High‑precision size‑based cell sorting
・Wide range of mesh sizes (5, 10, 15, 20, 100, 200 µm)
・Sterile
Catalog No. NC3918263 Supplier MURATA VIOS INC Supplier No. MZM1B015B50G
New approach to cell separation by size. Ideal for a wide range of cell processing applications, including fractionation, concentration, and filtration.
Highlights:
・Fast and simple processing
・High‑precision size‑based cell sorting
・Wide range of mesh sizes (5, 10, 15, 20, 100, 200 µm)
・Sterile