Learn More
Description
- Crystal Violet is taken up equally well by both gram-positive and negative bacterial cell walls. A crystal violet-iodine complex is formed within the peptidoglycan layer of the cell wall of each type of bacteria upon addition of iodine
- The advanced system helps to guard against accidental over-decolorization, thus reducing the possibility of mistaking a gram-positive organism for a gram-negative one
Specifications
Specifications
| Format | Liquid |
| Color | Violet |
| Volume (Metric) | 250 mL |
| Compatible Cells | Gram-negative and gram-positive bacteria |
| Label or Dye | Dye |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
