Learn More
Anton Paar Advanced density measurement of powders, solids and semi-solidsPowderProtect: no instrument contaminationBuilt-in Peltier control – no external bath requiredAccurate results for samples from 4.5 cm3 to 135 cm3