Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Advanced glycation end products | 1 MG
SDP

Catalog No. 5000426129
Encompass_Preferred

Advanced glycation end products (AGEs) are stable compounds formed by non-enzymatic reactions of reducing sugars with proteins, lipids, or nucleic acids. They are frequently utilized as a target for assessing the inhibitory effects of anti-glycosylation compounds.

  • Used for research purposes
  • Supplied as a liquid
  • Light yellow to yellow appearance
  • Concentration of 50 mg/mL
  • Formulated in 0.01 M PBS, pH 7.4
  • Store at -20°C for 2 years
  • Not classified as a hazardous substance or mixture

Catalog No. 50-004-26129 Supplier Medchemexpress LLC Supplier No. HYNP1651MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Advanced glycation end products (AGEs) are stable compounds formed by non-enzymatic reactions of reducing sugars with proteins, lipids, or nucleic acids. They are frequently utilized as a target for assessing the inhibitory effects of anti-glycosylation compounds.

  • Used for research purposes
  • Supplied as a liquid
  • Light yellow to yellow appearance
  • Concentration of 50 mg/mL
  • Formulated in 0.01 M PBS, pH 7.4
  • Store at -20°C for 2 years
  • Not classified as a hazardous substance or mixture

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.