Learn More
Hardy Diagnostics™ Advanced Grams Counterstain
SDP
Catalog No. 23001766 Shop All Hardy Diagnostics Products
Change view
Click to view available options
Quantity:
3.7 L
Catalog No. Quantity
23-001-766 3.7 L
1 options
This item is not returnable. View return policy
Catalog No. 23-001-766 Supplier Hardy Diagnostics™ Supplier No. S128A
Add to Cart
Add to Cart
This item is not returnable. View return policy
Use Advanced Counterstain to stain gram negatives a vivid and distinct red. The gram negative organisms will appear brighter with better definition at the edges
  • Advanced Counterstain™, a stronger counterstain than Safranin, will stain the gram-negative organisms a very deep red
  • Contrast between gram-positive organisms and gram-negative organisms in a mixed field is greatly enhanced
  • The advanced system helps to guard against accidental over-decolorization, thus reducing the possibility of mistaking a gram-positive organism for a gram-negative one
Specifications
Quantity 3.7 L
Color Red
Volume (Metric) 3.7 L