Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PURELL™ Advanced Hand Sanitizer Green Certified Foam
GREENER_CHOICESDP

Catalog No. 19044006 Shop All Gojo Industries, Inc. Products
Change view
Click to view available options
Mounting:
Wall Mount
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting
19-044-006 Wall Mount
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-044-006 Supplier PURELL™ Supplier No. 785102
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Refill for PURELL™ CS8 touch-free hand sanitizer dispensers.

  • Refill for PURELL™ CS8 touch-free hand sanitizer dispensers
  • Green Certified Foam
TRUSTED_SUSTAINABILITY

Specifications

Product Type Wall-mounted Dispenser
Color Clear
Length (Metric) 5.18 cm
Width (Metric) 3.45 cm
Height (Metric) 7.3 cm
Holds Hand Soap
Material Plastic
Mounting Wall Mount
No. of Compartments 1
Product Style Front-loading with Opening
For Use With (Equipment) PURELL™ CS8 Touch-Free Hand Sanitizer Dispensers
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.