Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Purell™ Advanced Hand Sanitizer Green Certified Gel
SDP

Catalog No. 19043813 Shop All Gojo Industries, Inc. Products
Change view
Click to view available options
Volume (English):
8 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
19-043-813 8 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-043-813 Supplier Gojo Industries Inc Supplier No. 969112
This item is not returnable. View return policy

Purell™ Advanced Hand Sanitizer Green Certified Gel is a table top pump bottle of refreshing gel made with naturally-renewable ethanol.

  • Refreshing gel made with naturally-renewable ethanol
  • Kills more than 99.99% of most common germs that may cause illness
  • Meets EcoLogo™ hand sanitizer standard for environmental leadership and proven performance
  • Meets the biobased standard for the USDA BioPreferred program
  • Outperforms other hand sanitizers ounce-for-ounce
  • Clinically-proven to maintain skin health
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Certifications/Compliance Meets UL EcoLogo Standard for Hand Sanitizer UL 2783, Meets USDA biobased Standard for hand sanitizer
For Use With (Application) Hand Sanitizer
Quantity 12 Pk.
Volume (English) 8 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.