Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PURELL™ Advanced Hand Sanitizer ULTRA NOURISHING™ Foam PROMO
SDP

Catalog No. 19043363W25 Shop All Gojo Industries, Inc. Products
Change view
Click to view available options
PURELL™ ULTRA NOURISHING™ Hand Sanitizer
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-043-363W25 PURELL™ ULTRA NOURISHING™ Hand Sanitizer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-043-363W25 Supplier PURELL™ Supplier No. PI835702

Refill (1200 mL) for PURELL™ ES10 touch-free hand sanitizer dispensers.

  • Enhanced with a moisturizer and antioxidant blend.
  • Gentle formulation clings to hands for mess-free sanitizing.
  • Contains vitamin B3 and is free of fragrance, dyes, parabens, phthalates, sulfates and harsh preservatives.
  • Made with our high-quality ethyl alcohol, produced in the USA.
  • Manufactured according to strict FDA safety and quality standards.
  • Refill bottles are easily recyclable to align with your sustainability standards.
  • Formula meets USDA Certified Biobased Product Standards for hand cleaners, ECOLOGO UL 2783, and Cradle to Cradle Certified
  • 1200 mL refill for the PURELL ES10 Hand Sanitizer Automatic Dispenser (sold separately) bottle is made with recyclable plastic components.
  • Collar and alkaline AA battery easily pull apart for easy recyclability and waste reduction.
  • Case Includes: 2-PURELL Advanced Hand Sanitizer ULTRA NOURISHING Foam, 1200 mL Refills for the ES10 Automatic Hand Sanitizer Dispenser.
  • ES10 dispensers (8324-E1, 8320-E1, 8328-E1) sold separately.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Hand Sanitizer
Color Clear
Form Foam
Size 1200 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.