Learn More
3B Scientific™ Advanced Manual Inflate Large Cuff Blood Pressure Monitor
SDPSCI_ED
Catalog No. s37585 Shop All 3B Scientific Products
Change view
Click to view available options
Size:
Large
Catalog No. Size
S37585 Large
1 options
This item is intended for educational purposes only.
Catalog No. S37585 Supplier 3B Scientific™ Supplier No. W64601L
Add to Cart
Add to Cart
This item is intended for educational purposes only.
Fast and accurate blood pressure measurement. The 3B Scientific Advanced Manual Inflate Blood Monitor is equipped with a Pressure Rating Indicator and Irregular Heartbeat feature.
  • Offers two innovative features, the Pressure Rating Indicator™ and Irregular Heartbeat feature
  • Pressure Rating Indicator provides a snapshot of your blood pressure classification based on your measurements and eliminates the guesswor
Specifications
Battery Type AA
Size Large
For Use With (Application) Montoring blood pressure
Includes On AA battery, carry case, four-week blood pressure logbook and quick start card in English, French and Spanish
Latex Free Yes
Product Type Blood presssure monitor