Learn More
Anton Paar Advanced monomode microwave reactor (850 W) for up to 300 °C and 30 bar. Compatible with borosilicate or SiC vials. Features touchscreen, stirring, pressure and IR sensors, and supports fiber-optic Raman probes. Includes USB/Ether
Catalog No. 5000580472
Encompass_Preferred
Power supply: 230V, 50 Hz/60 Hz, 1600 VA
Included:
- Spill bin (83740)
- 5 pcs. Reaction Vial G10
- 2 pcs. Reaction vial G30
- 7 pcs. Snap cap
- 20 pcs. Silicon septum
- 10 pcs. Stir bar PTFE 10x3
- Blind pin (89189)
- Adaptor ring (88999)
- 2 pcs. Cavity collar (83212)
- 3 pcs. Filter disc (84294)
- Piston cover key (89196)
Catalog No. 50-005-80472 Supplier Anton Paar Supplier No. 241699
Please contact your sales representative for a quote or further ordering instructions.
Power supply: 230V, 50 Hz/60 Hz, 1600 VA
Included:
- Spill bin (83740)
- 5 pcs. Reaction Vial G10
- 2 pcs. Reaction vial G30
- 7 pcs. Snap cap
- 20 pcs. Silicon septum
- 10 pcs. Stir bar PTFE 10x3
- Blind pin (89189)
- Adaptor ring (88999)
- 2 pcs. Cavity collar (83212)
- 3 pcs. Filter disc (84294)
- Piston cover key (89196)