Learn More
METTLER TOLEDO Advanced MR Analytical Balance
GREENER_CHOICE
Catalog No. 01804284 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Capacity:
120 g
220 g
320 g
Model:
MR104
MR204
MR304
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity Model
01-804-284 120 g MR104
01-804-282 220 g MR204
01-804-280 320 g MR304
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-804-284 Supplier METTLER TOLEDO Supplier No. 30666203
  • PRODUCT PROMOTION
    $5,856.50
    $3,459.00 / Each of 1
    Save $2,397.50
    41% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart
Request a Quote
METTLER TOLEDO Advanced MR Analytical Balances offer a highly precise weigh cell with FACT automatic internal adjustment.
User management within the 4.5-inch touchscreen increases efficiency of daily tasks with 8 built-in applications and statistical data analysis. Designed with full metal housing and overload protection, these balances will provide high quality performance for years to come. When your balance is not in use, enable the power-saving mode to reduce energy consumption and save costs.
MR Analytical Balances include:
  • FACT automatic internal adjustment
  • 4.5 inch color touchscreen
  • 8 built-in applications
  • Statistical data analysis
  • User management
  • Guided leveling
  • 4 interfaces: USB-A, USB-C, RS232, Ethernet
  • Bluetooth option
  • Full metal housing
  • Overload protection
See how it ranks among our balance lines:
  • XPR Excellence Analytical Balances
  • XPR-E Essential Analytical Balances
  • MX Advanced Analytical Balances
  • MR Advanced Analytical Balances
  • MA Standard Analytical Balances
  • FE Standard Analytical Balances
  • LA Standard Analytical Balances
TRUSTED_SUSTAINABILITY
Specifications
Connection Type USB-A, USB-C, RS232, Ethernet
Capacity 120 g
Display Type Color Touchscreen
Readability 0.1 mg
Material Metal
Model MR104
Stabilization Time 2 sec.
Type Analytical Balance
Electrical Requirements 100/240 V, 50/60 Hz
Legal for Trade No