Learn More
METTLER TOLEDO Advanced MR Precision Balance PROMO
Catalog No. 01804276PM Shop All METTLER TOLEDO Products
Change view
Click to view available options
Capacity:
320 g
Catalog No. Capacity
01-804-276PM 320 g
1 options
Catalog No. 01-804-276PM Supplier METTLER TOLEDO Supplier No. 30666215PM
Feature a precise weigh cell and FACT automatic internal adjustment for highly accurate results.
  • The 4.5-inch touchscreen with intuitive user management provides easy access to 9 built-in applications and increases efficiency in your workflow.
  • Guided leveling, multiple connectivity options and easy to clean balance features reduces time spent on routine tasks.
  • Designed with IP43 protection and full metal housing to ensure long lasting performance.
  • When your balance is not in use, enable the power-saving mode to reduce energy consumption and save costs.
MR Precision Balances include:
  • FACT Internal Adjustment
  • 4.5 inch touchscreen
  • 9 built in applications
  • User management
  • Guided leveling
  • Statistical data analysis
  • 4 interfaces: USB-A, USB-C, RS232, Ethernet
  • Bluetooth option
  • Full metal housing
  • Overload protection
The MR Balance is the successor to the ML or ME-T Balance. See how it ranks among our balance lines:
  • XPR Excellence Precision Balances
  • MX Advanced Precision Balances
  • MR Advanced Precision Balances
  • MA Standard Precision Balances
  • ME Standard Precision Balances
  • LA Standard Precision Balances
TRUSTED_SUSTAINABILITY
Specifications
Connection Type USB-A, USB-C, RS232, Ethernet
Capacity 320 g
Display Type Color Touchscreen
Readability 1 mg
Material Metal
Model MR303
Stabilization Time 1.5 sec.
Type Precision Balance
Electrical Requirements 100/240 V, 50/60 Hz
Legal for Trade No