Description
- 9 W dimmable LED modules arranged in two semi-circular arrays with 11W total consumption, infinite dimming (1 to 100%), 5700 lux at 13 in. focal length, 900 lumens, 4000 K CCT, and CRI 80 for high-quality illumination.
- 5-diopter (2.25X) 5 in. diameter optical-quality glass lens with option to attach secondary 4-, 6-, or 10-diopter STAYS lens for increased magnification.
- Heavy-duty internal-spring 45 in. or 30 in. parallel, three-pivot K-arm for stable, precise, and flexible positioning during tasks.
- Supplied with cable and plug, featuring integrated auto shut-off after 4 or 9 hours for energy efficiency and safety.
- Standard mounting includes edge clamp or weighted base, with additional mounting options available for versatile installation.
- Durable white finish with optional matching handle accessory for improved usability and control.
Specifications
Specifications
| Type | Magnifier |
| Autoclavable | Not Autoclavable |
| Color | White |
| Sterility | Non-sterile |