Learn More
JDM LAB SOLUTIONS LLC Advanced Peltier Technology for rapid heat-up and cool down times. Temperature control for continuous operation. Wide variety of interchangeable plate blocks. Programmable time, temperature segments
Supplier:  JDM LAB SOLUTIONS LLC DBC05A
Encompass
Crystal Digital Heating/Cooling Dry Baths are ideal for a variety of Life Science and General Laboratory applications, featuring a wide temperature range, programmability and user-friendly LCD display. These baths offer innovative block construction and increased flexibility. Advanced Peltier technology ensures rapid and accurate temperature management, without a refrigeration system, and significantly reduce experiment time. Dry temperature control maintains a clean environment, reducing contamination risk. This vibration free system is ideal for incubations where stable conditions are crucial.
Catalog No. NC3933558