Learn More
JDM LAB SOLUTIONS LLC Advanced Peltier Technology for rapid heat-up and cool down times. Temperature control for continuous operation. Wide variety of interchangeable plate blocks. Programmable time, temperature segments
Catalog No. NC3933561
Encompass
Crystal Digital Heating/Cooling Dry Baths are ideal for a variety of Life Science and General Laboratory applications, featuring a wide temperature range, programmability and user-friendly LCD display. These baths offer innovative block construction and increased flexibility. Advanced Peltier technology ensures rapid and accurate temperature management, without a refrigeration system, and significantly reduce experiment time. Dry temperature control maintains a clean environment, reducing contamination risk. This vibration free system is ideal for incubations where stable conditions are crucial.
Catalog No. NC3933561 Supplier JDM LAB SOLUTIONS LLC Supplier No. DBC06A
Crystal Digital Heating/Cooling Dry Baths are ideal for a variety of Life Science and General Laboratory applications, featuring a wide temperature range, programmability and user-friendly LCD display. These baths offer innovative block construction and increased flexibility. Advanced Peltier technology ensures rapid and accurate temperature management, without a refrigeration system, and significantly reduce experiment time. Dry temperature control maintains a clean environment, reducing contamination risk. This vibration free system is ideal for incubations where stable conditions are crucial.