Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier™ Advanced Physics with Vernier — Beyond Mechanics
SDPSCI_ED

Catalog No. S16240ND Shop All Vernier Software Products
Change view
Click to view available options
Grade:
9 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S16240ND 9 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S16240ND Supplier Vernier™ Supplier No. PHYSABM
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Advanced Physics with Vernier — Beyond Mechanics

Use Vernier probeware to teach concepts at the college physics, AP* Physics, or IB** Physics level. Use it in addition to and for a comprehensive set of topics.

  • 22 student experiments complete with a materials list, instructions for data collection with Logger Pro 3 or LabQuest App, data tables, and questions
  • Access to up-to-date versions of the experiments
  • Instructor Information section for each experiment with complete directions for setting up experiments, helpful hints, sample graphs, and data
  • Word-processing files of the student experiments, so you can edit the files to match your teaching preferences
  • Electronic support material including videos
  • Printed copy of the book

Warranty and Services

5-year limited warranty

Specifications

Grade 9 to 12
Description ISBN 978-1-929075-90-4
Format Printed Lab Book
Languages English
For Use With (Application) Educational use only
Subject Physics
Author Larry Dukerich
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.