Learn More
Perkin Elmer US LLC Advanced Platform THGA Graphite Tubes, Pkg. 20
Catalog No. 502103720
Encompass_Preferred
The exclusive and patented Advanced Platform design offers an alternative to our Standard Integrated Platform design. With the current Integrated Platform design, the platform attaches to the tube wall along a single side that runs the length of the platform.
Catalog No. 50-210-3720 Supplier Perkin Elmer US LLC Supplier No. N9307831
The exclusive and patented Advanced Platform design offers an alternative to our Standard Integrated Platform design. With the current Integrated Platform design, the platform attaches to the tube wall along a single side that runs the length of the platform.