Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One Advanced TC Cell Culture Dishes
GREENER_CHOICE

Catalog No. 07000598 Shop All Greiner Bio-One Products
Change view
Click to view available options
Diameter (Metric):
100 mm
145 mm
Volume (Metric) Working:
15 to 17 mL
25 to 27 mL
Height (Metric):
20 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Volume (Metric) Working Height (Metric)
07-000-598 145 mm 25 to 27 mL 20 mm
07-000-686 100 mm 15 to 17 mL 20 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 07-000-598 Supplier Greiner Bio-One Supplier No. 639960
Add to Cart
Add to Cart

For cultivation of fastidious and sensitive cells. Greiner Bio-One Advanced TC Cell Culture Dishes are free of detectable DNase, RNase, human DNA.

  • Non-pyrogenic and non-cytotoxic
  • Optimised cell adherence
  • Increased cell yield
  • Homogenous cell growth
  • Higher transgene activity
  • Improved assay consistency
  • Non-biological surface, sterile
  • Shelf
TRUSTED_SUSTAINABILITY

Specifications

Closure Color Clear
Closure Included Closure Included
Closure Material Polystyrene
Bottom Shape Flat
Capacity (Metric) 240 mL
Color Clear
Culture Area 143 sq. cm
Material Polystyrene
Shape Round
Sterility Sterile
Type Cell Culture Dish
Volume (Metric) Working 25 to 27 mL
Includes Lid
Height (Metric) 20 mm
Diameter (Metric) 145 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.