Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One Advanced TC Treated Sterile Cell Culture Multi-well Plates
GREENER_CHOICE

Catalog No. 07000682 Shop All Greiner Bio-One Products
Change view
Click to view available options
Well Count:
12
24
48
Capacity (Metric):
1.7 mL
3.3 mL
6.5 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Well Count Capacity (Metric)
07-000-682 24 3.3 mL
07-000-691 12 6.5 mL
07-000-729 48 1.7 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 07-000-682 Supplier Greiner Bio-One Supplier No. 662960
Add to Cart
Add to Cart

For cultivation of fastidious cells. Grenier Bio-One Advanced TC Treated Sterile Cell Culture Multi-well Plates are free of detectable DNase, RNase, human DNA.

  • Provide conditions for improved cell adherence, accelerated cell expansion and increase overall cell yield
  • For use with serum reduced and serum-free media
  • Allows differentiation of semi-adherent cells
  • Non-pyrogenic and non-cytotoxic
  • Lid with condensation rings

Warranty and Services

No

TRUSTED_SUSTAINABILITY

Specifications

Closure Color Clear
Closure Included Closure Included
Closure Material Polystyrene
Autoclavable Not Autoclavable
Bottom Shape Flat
Capacity (Metric) 3.3 mL
Color Clear
Culture Area 1.9 sq. cm
Material Polystyrene
Shape Rectangular
Sterility Sterile
Surface Treatment Advanced TC
Type Cell Culture Dish
Well Count 24
Includes Lid
No. per Case 100
No. per Pack 1
Dimensions (L x W x H) 127.8 x 85.5 x 16.5 mm
Length (Metric) 127.76 mm
Width (Metric) 85.48 mm
Height (Metric) 19 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.