Learn More
Greiner Bio-One Advanced TC Treated Suspension Cell Culture Flasks with Standard Screw Cap
Catalog No. 07000766 Shop All Greiner Bio-One Products
Change view
Click to view available options
Culture Area:
175 cm2
25 cm2
75 cm2
Volume (Metric) Working:
50mL
250mL
550 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Culture Area Volume (Metric) Working
07-000-766 25 cm2 50mL
07-000-654 75 cm2 250mL
07-000-661 175 cm2 550 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 07-000-766 Supplier Greiner Bio-One Supplier No. 690960
For maintenance of adherent cells or suspension cell culture. Greiner Bio-One Advanced TC Treated Suspension Cell Culture Flasks with Standard Screw Cap provides optimal conditions for cultivation of sensitive and fastidious cells.
  • Made of high-grade polystyrene
  • Free of detectable DNases, RNases, human DNA, endotoxins and are non-cytotoxic
  • Stacking rim on the top of the flasks ensures firm standing and easy stackability in the incubator
  • Printed graduation for easier filling on both sides
  • Provides homogeneous and optimized cell growth
TRUSTED_SUSTAINABILITY
Specifications
Material Polystyrene
Surface Coating Advanced TC
Disposable Single-use
Graduated Yes
Sterility Sterile
Bottom Shape Flat
Closure Type Standard screw cap
Culture Area 25 cm2
Surface Treatment Tissue Culture-treated
Volume (Metric) Working 50mL