Learn More
Chemglass Life Sciences Advantaflex, 36" Extension Line, Plugged Ends, Sterile
SDP
Catalog No. 5000083212
Encompass
Fluid Transfer Extension Lines are produced in a Class 7 Cleanroom with weldable size 16 (1/8 x 1/4) TPE tubing. They are available in both Advantaflexand C-Flex tubing, both with plugged ends. The Fluid transfer lines are designed to aid in the process of transfer and filling of bioreactors, mixers and fermenters. Individually wrapped and sterile.
Catalog No. 50-000-83212 Supplier Chemglass Life Sciences Supplier No. CLS1390A036
Fluid Transfer Extension Lines are produced in a Class 7 Cleanroom with weldable size 16 (1/8 x 1/4) TPE tubing. They are available in both Advantaflexand C-Flex tubing, both with plugged ends. The Fluid transfer lines are designed to aid in the process of transfer and filling of bioreactors, mixers and fermenters. Individually wrapped and sterile.