Learn More
New Age Industries Inc ADVANTAFLEX SZ 16 TUBING, 500'
SDP
Catalog No. NC1494860
Encompass_Preferred
AdvantaFlex Size 16 Tubing, 500' spool Weldable, Autoclaveable, I.D. 0.125in, Wall 0.062in, O.D. 0.25in, PSI 30
Catalog No. NC1494860 Supplier New Age Industries Inc Supplier No. APAFBP01250250500S
AdvantaFlex Size 16 Tubing, 500' spool Weldable, Autoclaveable, I.D. 0.125in, Wall 0.062in, O.D. 0.25in, PSI 30