Learn More
Analytical Sales & Services Analytical Sales Adhesive Sealing Film, Teflon Material, 96-Well, Scored, 100 Per Package
Supplier:  Analytical Sales & Services 961802/DEL
Encompass
The Analytical Sales & Services Adhesive Sealing Film is designed for scientific applications, providing a firm seal on microplates to prevent cross-contamination and evaporation. Its ultra-thin Teflon (PTFE) construction and scoring enhance compatibility with PCR applications and automated liquid handling systems. This film ensures reliable performance with no needle clogging, outstanding solvent resistance, and a temperature range of -80ºC to +120ºC, making it an essential tool in labs.
Catalog No. NC0525923