Learn More
Accuflow Systems AdvantaPure Platinum-Cured Silicone Tubing, 0.250 in ID, 0.375 in OD
Catalog No. NC3872068
Encompass_Preferred
AdvantaSil APST is a platinum-cured, translucent silicone tubing produced in a Class 7 clean room for critical pharmaceutical, biopharmaceutical, food, and cosmetic fluid-transfer applications. The unreinforced construction provides flexibility for single-use and overmolding applications while meeting biocompatibility and extractables standards.
  • Translucent platinum-cured silicone, unreinforced.
  • 0.250 in inner diameter, 0.375 in outer diameter, 0.063 in wall.
  • 100 ft coil length.
  • 50 Shore A hardness.
  • Temperature range -100°F to 400°F (-73.3°C to 204.4°C).
  • Sterilizable by autoclave or gamma irradiation; validated sterility available.
  • Meets USP Class VI, FDA CFR 177.2600, ISO 10993, NSF 51, and European Pharmacopoeia 3.1.9.
  • Produced in a Class 7 clean room; particulate testing per USP 788.
Catalog No. NC3872068 Supplier Accuflow Systems Supplier No. APST02500375K
AdvantaSil APST is a platinum-cured, translucent silicone tubing produced in a Class 7 clean room for critical pharmaceutical, biopharmaceutical, food, and cosmetic fluid-transfer applications. The unreinforced construction provides flexibility for single-use and overmolding applications while meeting biocompatibility and extractables standards.
  • Translucent platinum-cured silicone, unreinforced.
  • 0.250 in inner diameter, 0.375 in outer diameter, 0.063 in wall.
  • 100 ft coil length.
  • 50 Shore A hardness.
  • Temperature range -100°F to 400°F (-73.3°C to 204.4°C).
  • Sterilizable by autoclave or gamma irradiation; validated sterility available.
  • Meets USP Class VI, FDA CFR 177.2600, ISO 10993, NSF 51, and European Pharmacopoeia 3.1.9.
  • Produced in a Class 7 clean room; particulate testing per USP 788.