Learn More
Accuflow Systems AdvantaPure Silicone Tubing, Platinum Cured, 0.375 in ID, 150 ft
Catalog No. NC3872069
Encompass_Preferred
AdvantaSil APST platinum-cured silicone tubing is a low-volatile, unreinforced tubing produced in a cleanroom for critical pharmaceutical, biopharm, cosmetic, and food applications. It is designed for single-use fluid transfer in regulated processes and has undergone standardized extractables and biocompatibility testing. Not intended for implantation, continuous steam, or elevated pressure applications.
  • Platinum-cured silicone for low volatile content and biocompatibility.
  • Inner diameter 0.375 in, outer diameter 0.625 in, wall thickness 0.125 in.
  • 150 ft spool for extended continuous runs and fewer joins.
  • Temperature range -50° F to 400° F (-45° C to 204.4° C).
  • Meets USP Class VI, FDA CFR 177.2600, ISO 10993, NSF 51, and European Pharmacopoeia 3.1.9.
  • Produced and packaged in ISO cleanroom conditions for regulated manufacturing.
Catalog No. NC3872069 Supplier Accuflow Systems Supplier No. APST03750625150S
AdvantaSil APST platinum-cured silicone tubing is a low-volatile, unreinforced tubing produced in a cleanroom for critical pharmaceutical, biopharm, cosmetic, and food applications. It is designed for single-use fluid transfer in regulated processes and has undergone standardized extractables and biocompatibility testing. Not intended for implantation, continuous steam, or elevated pressure applications.
  • Platinum-cured silicone for low volatile content and biocompatibility.
  • Inner diameter 0.375 in, outer diameter 0.625 in, wall thickness 0.125 in.
  • 150 ft spool for extended continuous runs and fewer joins.
  • Temperature range -50° F to 400° F (-45° C to 204.4° C).
  • Meets USP Class VI, FDA CFR 177.2600, ISO 10993, NSF 51, and European Pharmacopoeia 3.1.9.
  • Produced and packaged in ISO cleanroom conditions for regulated manufacturing.