Learn More
Advantec MFS, Inc 47 mm Glass Fiber Filter, 100/pk
Supplier:  Advantec MFS, Inc GF7547MM
Encompass_Preferred
Nominal Retention (um): 0.3. Thickness (mm): 0.35. Collection Efficiency % (0.3 um POA): 99.999.
Catalog No. NC0513025