Learn More
Cole-Parmer Advantec N05C7.0CM Grade 5C Ashless Quantitative Filters, <5 um, 7 cm; 100/Pk
Supplier:  Cole-Parmer 8105144
Encompass_Preferred
Ideal for gravimetric analysis and environmental monitoring. These papers are made of the highest quality alpha cotton cellulose with low ash content. Papers are double acid washed in hydrochloric acid followed by hydroflouric acid, then rinsed with ultrapure water to neutralize. Sold in packs of 100.
Catalog No. NC0327835