Learn More
Cole-Parmer Advantec N051A200X200MM Paper, blotting and chromatography; Grade 51A, 200 x 200 mm; pack of 100 sheets
Catalog No. NC1540573 Shop All Cole-Parmer Products
Encompass_Preferred
These papers are carefully tested for spot formation, capillary action, water flow rate, and absorption speed to ensure performance uniformity and repeatability. Applications include chromatography, electrophoresis/blotting, and separation of heavily loaded solutes.
Catalog No. NC1540573 Supplier Cole-Parmer Supplier No. 2850170
These papers are carefully tested for spot formation, capillary action, water flow rate, and absorption speed to ensure performance uniformity and repeatability. Applications include chromatography, electrophoresis/blotting, and separation of heavily loaded solutes.