Learn More
Enterprise Technology Solutions Advantus Corp. ID Cards, Laminated PVC, 2-/18"x3-3/8", 100/PK, White
SDP
Catalog No. 5000832593
Encompass_Preferred
Make your own professional ID cards, employee badges and security passes. Each blank card is constructed of durable, thick PVC. Cards are laminated for dye sublimation and thermal (up to 172 degrees) printing. Each card measures 2-1/8" x 3-3/8".
Catalog No. 50-008-32593 Supplier Enterprise Technology Solutions Supplier No. AVT97034
Make your own professional ID cards, employee badges and security passes. Each blank card is constructed of durable, thick PVC. Cards are laminated for dye sublimation and thermal (up to 172 degrees) printing. Each card measures 2-1/8" x 3-3/8".