Learn More
Enterprise Technology Solutions Advantus Corp. Lanyards, Breakaway, w/Plastic Hook, 36"L, 12/PK, Black
SDP
Catalog No. 5000832412
Encompass_Preferred
These durable and lightweight breakaway lanyards are designed with safety in mind. Each lanyard is designed with a breakaway feature that ensures your safety if the lanyard is snagged, pulled or caught. It is equipped with a black plastic hook for attaching identification cards or name badges and ideal for use in non-metal environments such as security areas of airports and CAT or MRI areas of hospitals. Lanyard measures 36" in length.
Catalog No. 50-008-32412 Supplier Enterprise Technology Solutions Supplier No. AVT97131
These durable and lightweight breakaway lanyards are designed with safety in mind. Each lanyard is designed with a breakaway feature that ensures your safety if the lanyard is snagged, pulled or caught. It is equipped with a black plastic hook for attaching identification cards or name badges and ideal for use in non-metal environments such as security areas of airports and CAT or MRI areas of hospitals. Lanyard measures 36" in length.