Learn More
Biotium ADVASEP-7
Catalog No. NC1669433
Encompass_Preferred
ADVASEP-7 is a sulfonated β-cyclodextrin derivative that has been reported to reduce background fluorescence when using SynaptoGreen C4 to stain brain slices. Properties: Average MW: 2163; C42H70-nO35(C4H8SO3Na)n•hydrate where n = Average Degree of Substitution 6.5; White solid soluble in water; Store at 4°C
Catalog No. NC1669433 Supplier Biotium Supplier No. 70029
ADVASEP-7 is a sulfonated β-cyclodextrin derivative that has been reported to reduce background fluorescence when using SynaptoGreen C4 to stain brain slices. Properties: Average MW: 2163; C42H70-nO35(C4H8SO3Na)n•hydrate where n = Average Degree of Substitution 6.5; White solid soluble in water; Store at 4°C