Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OHAUS™ Adventurer™ Precision Balances
SCI_ED

Catalog No. S28593 Shop All OHAUS Products
Change view
Click to view available options
Model:
AX1502
AX223
AX4201
AX423
AX5202
AX523
AX622
AX8201
Capacity:
220 g
420 g
520 g
620 g
1.5 kg
4.2 kg
5.2 kg
8.2 kg
Readability:
0.001 g
0.01 g
0.1 g
Stabilization Time:
≤1.5 sec.
≤2 sec.
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Capacity Readability Stabilization Time
S28593 AX5202 5.2 kg 0.01 g ≤1.5 sec.
S28577 AX1502 1.5 kg 0.01 g ≤1.5 sec.
S28581 AX223 220 g 0.001 g ≤2 sec.
S28586 AX4201 4.2 kg 0.1 g ≤1.5 sec.
S28589 AX423 420 g 0.001 g ≤2 sec.
S28594 AX523 520 g 0.001 g ≤2 sec.
S28596 AX622 620 g 0.01 g ≤1.5 sec.
S28598 AX8201 8.2 kg 0.1 g ≤1.5 sec.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. S28593 Supplier OHAUS™ Supplier No. 30100617/EMD
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Strikes the ideal balance between inventive features and functional, uncomplicated weighing capabilities. OHAUS Adventurer Precision Balances deliver stable and reliable weighing results for routine weighing tasks. With fast stabilization times, these balances improve productivity in the laboratory.

  • External Calibration: traditional calibration in which the operator manually calibrates the balance with their choice of calibration weight value to ensure accuracy available on every model
  • AutoCal™: select models feature OHAUS automatic internal calibration system that performs routine maintenance by calibrating the balance daily
  • Color touchscreen offers easy and fast operation of Adventurer applications
  • Operate and access nine application modes and abundant features that eliminate the need to do several manual calculations through the modern color touchscreen
  • Operators can wear laboratory gloves while utilizing the touchscreen, eliminating the inconvenience and hazards associated with constantly putting on and removing gloves
  • In addition to the touchscreen, Adventurer also has six mechanical keys that provide tactile feedback and allow the operator to perform repetitive operations such as tare, zero, calibration, and print
  • Application modes: weighing, dynamic weighing, display hold, parts counting, density determination, totalization, percent weighing, check weighing, formulation
  • Real-time clock function keeps accurate time even during power loss and the GLP data capability has the ability to record Sample name, Project names and Balance IDs to help meet traceability and compliance requirements
  • Cloning feature allows you to save user and application settings to a USB flash drive which can be easily used to configure additional Adventurer balances
  • When using the minimum weight feature, the display clearly indicates that your current sample weight is below your defined minimum limit; simply increase your sample weight to assure that your results are up to your standards
  • Space-saving draftshield designed to improve user experience and accessibility
  • Additional features include: RS232 interface, integral weigh below hook, full housing in-use cover, removable stainless steel pan, die-cast metal bottom housing, security bracket, illuminated up-front level indicator, four adjustable feet, software lockout menus, stability indicator, software overload/underload indicators, user selectable environmental settings, audible indicator, user selectable brightness settings, auto dim, touchscreen calibration, auto tare, user selectable operating language (nine)

Dual USB Ports

  • A front USB host port is easily accessible and makes it simple to load data from the balance on to a flash drive without having to reach around to the back or move the balance
  • A second USB device is located at the rear of the balance that can be used to connect the balance to a PC
  • The connectivity options help meet traceability requirements in traditional installations

Compliance

Product Safety: CSA C22.2 No. 61010-1, UL 61010-1, IEC 61010-1. Electromagnetic Compatibility: FCC Part 15 Class A, ICES-001 Class A, IEC 61326-1 (emissions Class B, immunity Industrial requirements)

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Product Safety: CSA C22.2 No. 61010-1, UL 61010-1,
Connection Type RS-232, USB device, USB host
Capacity 5.2 kg
Display Type Touchscreen
Readability 0.01 g
For Use With (Application) Weighing, Parts Counting, Percent Weighing, Check
Includes Not Applicable
Linearity ±0.02 g
Material Metal Base, ABS Top Housing, Stainless Steel Pan
Model AX5202
Repeatability ±0.01 g
Stabilization Time ≤1.5 sec.
Temperature Range (English) 50°F to 86°F
Temperature Range (Metric) 10°C to 30°C
Type Precision Balance, Internal Calibration
Width (Metric) Pan 195 mm
Depth (Metric) Pan 175 mm
Diameter (Metric) Outer Pan Not Applicable
Weight (English) 10.2 lb.
Weight (Metric) 4.6 kg
Electrical Requirements AC Adapter Input: 100-240V 0.3A 50-60Hz; AC Adapte
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.