Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OHAUS™ Adventurer™ Precision Balances with External Calibration

Catalog No. 01919406 Shop All OHAUS Products
Change view
Click to view available options
Model:
AX1502/E
AX1502N/E
AX2201/E
AX2202/E
AX2202N/E
AX223N/E
AX4201/E
AX4202/E
AX4202N/E
AX422/E
AX423/E
AX423N/E
AX523/E
AX523N/E
AX622N/E
AX8201/E
AX8201N/E
AX822/E
Capacity:
220 g
420 g
520 g
620 g
820 g
1.5 kg
2.2 kg
4.2 kg
8.2 kg
Readability:
0.001 g
0.001, 0.01 g (Certified)
0.01 g
0.1 g
Stabilization Time:
≤1.5 sec.
≤2 sec.
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Capacity Readability Stabilization Time
01-919-406 AX2201/E 2.2 kg 0.1 g ≤1.5 sec.
01-920-260 AX622N/E 620 g 0.01 g ≤1.5 sec.
01-920-263 AX4202N/E 4.2 kg 0.01 g ≤1.5 sec.
01-920-261 AX1502N/E 1.5 kg 0.01 g ≤1.5 sec.
01-920-264 AX8201N/E 8.2 kg 0.1 g ≤1.5 sec.
01-920-262 AX2202N/E 2.2 kg 0.01 g ≤1.5 sec.
01-920-276 AX423/E 420 g 0.001 g ≤2 sec.
01-920-282 AX423N/E 420 g 0.01 g ≤2 sec.
01-920-280 AX223N/E 220 g 0.001, 0.01 g (Certified) ≤2 sec.
01-920-283 AX523N/E 520 g 0.01 g ≤2 sec.
01-919-340 AX4201/E 4.2 kg 0.1 g ≤1.5 sec.
01-919-342 AX8201/E 8.2 kg 0.1 g ≤1.5 sec.
01-919-351 AX1502/E 1.5 kg 0.01 g ≤1.5 sec.
01-919-353 AX2202/E 2.2 kg 0.01 g ≤1.5 sec.
01-919-355 AX4202/E 4.2 kg 0.01 g ≤1.5 sec.
01-919-362 AX523/E 520 g 0.001 g ≤2 sec.
01-919-404 AX422/E 420 g 0.01 g ≤1.5 sec.
01-919-405 AX822/E 820 g 0.01 g ≤1.5 sec.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
Catalog No. 01-919-406 Supplier OHAUS™ Supplier No. 30248912
Only null left
Add to Cart
Add to Cart

Strikes the ideal balance between inventive features and functional, uncomplicated weighing capabilities.

Delivers stable and reliable weighing results for routine weighing tasks.

  • Fast stabilization time improves productivity in the laboratory
  • External Calibration: traditional calibration in which the operator manually calibrates the balance with their choice of calibration weight value to ensure accuracy available on every model
  • Color touchscreen offers easy and fast operation of Adventurer applications
  • Operate and access nine application modes and abundant features that eliminate the need to do several manual calculations through the modern color touchscreen
  • Operators can wear laboratory gloves while utilizing the touchscreen, eliminating the inconvenience and hazards associated with constantly putting on and removing gloves
  • In addition to the touchscreen, Adventurer also has six mechanical keys that provide tactile feedback and allow the operator to perform repetitive operations such as tare, zero, calibration, and print
  • Application modes: weighing, dynamic weighing, display hold, parts counting, density determination, totalization, percent weighing, check weighing, formulation
  • Real-time clock function keeps accurate time even during power loss and the GLP data capability has the ability to record Sample name, Project names and Balance IDs to help meet traceability and compliance requirements
  • Cloning feature allows you to save user and application settings to a USB flash drive which can be easily used to configure additional Adventurer balances
  • When using the minimum weight feature, the display clearly indicates that your current sample weight is below your defined minimum limit; simply increase your sample weight to assure that your results are up to your standards
  • Space-saving draftshield designed to improve user experience and accessibility for some models
  • Additional features include: RS232 interface, integral weigh below hook, full housing in-use cover, removable stainless steel pan, die-cast metal bottom housing, security bracket, illuminated up-front level indicator, four adjustable feet, software lockout menus, stability indicator, software overload/underload indicators, user selectable environmental settings, audible indicator, user selectable brightness settings, auto dim, touchscreen calibration, auto tare, user selectable operating language (nine)

Dual USB Ports

  • A front USB host port is easily accessible and makes it simple to load data from the balance on to a flash drive without having to reach around to the back or move the balance
  • A second USB device is located at the rear of the balance that can be used to connect the balance to a PC
  • The connectivity options help meet traceability requirements in traditional installations

Compliance

Product Safety: CSA C22.2 No. 61010-1, UL 61010-1, IEC 61010-1. Electromagnetic Compatibility: FCC Part 15 Class A, ICES-001 Class A, IEC 61326-1 (emissions Class B, immunity Industrial requirements)

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Product Safety: CSA C22.2 No. 61010-1, UL 61010-1,
Connection Type RS-232, USB device, USB host
Capacity 2.2 kg
Display Type Touchscreen
Readability 0.1 g
For Use With (Application) Weighing, Parts Counting, Percent Weighing, Check
Linearity ±0.2 g
Material Metal Base, ABS Top Housing, Stainless Steel Pan
Model AX2201/E
Repeatability ±0.1 g
Stabilization Time ≤1.5 sec.
Temperature Range (English) 50°F to 86°F
Temperature Range (Metric) 10°C to 30°C
Type Precision Balance
Width (Metric) Pan 175 mm
Height (Metric) Pan 175 mm
Depth (Metric) Pan 195 mm
Weight (English) 10.2 lb.
Weight (Metric) 4.6 kg
Electrical Requirements AC Adapter Input: 100-240V 0.3A 50-60Hz; AC Adapte
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.